RIE156330755 Rieke Metals, Inc.

2-(1-Oxo-2-phenylethyl)benzoic acid

  • MDL number MFCD00189277
  • Cas Number : 33148-55-9
  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7101

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7101
Availability details for
Quantity
  • Availability
  • Add to Cart