RIE156330757 Rieke Metals, Inc.

Ethyl 4-[4-(N,N-Dimethylamino)phenyl]-4-oxobutanoate

  • MDL number MFCD09801904
  • Cas Number : 117937-10-7
  • Molecular Weight : 249.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7700B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7700B
Availability details for
Quantity
  • Availability
  • Add to Cart