RIE156330760 Rieke Metals, Inc.

4-(2,3,4-trimethoxyphenyl)-4-oxobutyric acid

  • MDL number MFCD09801784
  • Cas Number : 63213-41-2
  • Molecular Weight : 268.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7213B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7213B
Availability details for
Quantity
  • Availability
  • Add to Cart