RIE156330761 Rieke Metals, Inc.

5-(5-Chloro-2-thienyl)-3-methyl-5-oxovaleric acid

  • MDL number MFCD06201679
  • Cas Number : 845781-52-4
  • Molecular Weight : 246.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7292

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7292
Availability details for
Quantity
  • Availability
  • Add to Cart