RIE156330763 Rieke Metals, Inc.

4-oxo-4-(4-n-pentylphenyl)butyric acid

  • MDL number MFCD00173763
  • Cas Number : 64779-07-3
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7151B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7151B
Availability details for
Quantity
  • Availability
  • Add to Cart