RIE156330765 Rieke Metals, Inc.

3-Propionylbenzoic acid

  • MDL number MFCD02260860
  • Cas Number : 74380-71-5
  • Molecular Weight : 178.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7106

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7106
Availability details for
Quantity
  • Availability
  • Add to Cart