RIE156330766 Rieke Metals, Inc.

4-(4-n-Butylphenyl)-4-oxobutyric acid

  • MDL number MFCD00173764
  • Cas Number : 72271-71-7
  • Molecular Weight : 234.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7214B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7214B
Availability details for
Quantity
  • Availability
  • Add to Cart