RIE156330767 Rieke Metals, Inc.

5-(4-n-Butylphenyl)-5-oxovaleric acid

  • MDL number MFCD02260957
  • Cas Number : 97678-95-0
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7214C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7214C
Availability details for
Quantity
  • Availability
  • Add to Cart