RIE156330770 Rieke Metals, Inc.

ethyl 6-(3-chlorophenyl)-6-oxohexanoate

  • MDL number MFCD01320238
  • Cas Number : 333355-35-4
  • Molecular Weight : 268.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7544D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7544D
Availability details for
Quantity
  • Availability
  • Add to Cart