RIE156330771 Rieke Metals, Inc.

4-(4-Hexylphenyl)-4-oxobutyric acid

  • MDL number MFCD00173762
  • Cas Number : 64779-08-4
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7152B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7152B
Availability details for
Quantity
  • Availability
  • Add to Cart