RIE156330777 Rieke Metals, Inc.

Ethyl 5-methyl-4-oxohexanoate

  • MDL number MFCD02261303
  • Cas Number : 54857-48-6
  • Molecular Weight : 172.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7631B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7631B
Availability details for
Quantity
  • Availability
  • Add to Cart