RIE156330781 Rieke Metals, Inc.

4-(2-Ethoxyphenyl)-4-oxobutyric acid

  • MDL number MFCD02260978
  • Cas Number : 39595-35-2
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7154B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7154B
Availability details for
Quantity
  • Availability
  • Add to Cart