RIE156330782 Rieke Metals, Inc.

4-[4-(Ethylthio)phenyl]-4-oxobutyric acid

  • MDL number MFCD06201686
  • Cas Number : 7028-68-4
  • Molecular Weight : 238.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7217B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7217B
Availability details for
Quantity
  • Availability
  • Add to Cart