RIE156330783 Rieke Metals, Inc.

5-[4-(Ethylthio)phenyl]-5-oxovaleric acid

  • MDL number MFCD06201687
  • Cas Number : 845790-39-8
  • Molecular Weight : 252.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7217C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7217C
Availability details for
Quantity
  • Availability
  • Add to Cart