RIE156330786 Rieke Metals, Inc.

5-(3,4-Dimethoxyphenyl)-3-methyl-5-oxovaleric acid

  • MDL number MFCD06201680
  • Cas Number : 106591-88-2
  • Molecular Weight : 266.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7315

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7315
Availability details for
Quantity
  • Availability
  • Add to Cart