RIE156330787 Rieke Metals, Inc.

4-Oxononanoic acid

  • MDL number MFCD00138072
  • Cas Number : 6064-52-4
  • Molecular Weight : 172.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7089B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7089B
Availability details for
Quantity
  • Availability
  • Add to Cart