RIE156330788 Rieke Metals, Inc.

5-oxodecanoic acid

  • MDL number MFCD01320165
  • Cas Number : 624-01-1
  • Molecular Weight : 186.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7089C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7089C
Availability details for
Quantity
  • Availability
  • Add to Cart