RIE156330789 Rieke Metals, Inc.

4-(5-Chloro-2-thienyl)-4-oxobutyric acid

  • MDL number MFCD03964581
  • Cas Number : 70685-06-2
  • Molecular Weight : 218.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7220B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7220B
Availability details for
Quantity
  • Availability
  • Add to Cart