RIE156330790 Rieke Metals, Inc.

5-(5-Chloro-2-thienyl)-5-oxovaleric acid

  • MDL number MFCD06201688
  • Cas Number : 845790-40-1
  • Molecular Weight : 232.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7220C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7220C
Availability details for
Quantity
  • Availability
  • Add to Cart