RIE156330792 Rieke Metals, Inc.

3-Methyl-5-oxo-5-phenylvaleric acid

  • MDL number MFCD01896657
  • Cas Number : 2840-61-1
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7116

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7116
Availability details for
Quantity
  • Availability
  • Add to Cart