RIE156330793 Rieke Metals, Inc.

4-[3,4-(Methylenedioxy)phenyl]-4-oxobutyric acid

  • MDL number MFCD00508396
  • Cas Number : 41764-07-2
  • Molecular Weight : 222.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7222B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7222B
Availability details for
Quantity
  • Availability
  • Add to Cart