RIE156330797 Rieke Metals, Inc.

Ethyl 4-(3-nitrophenyl)-4-oxobutyrate

  • MDL number MFCD02261329
  • Cas Number : 75227-18-8
  • Molecular Weight : 251.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7637B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7637B
Availability details for
Quantity
  • Availability
  • Add to Cart