RIE156330798 Rieke Metals, Inc.

ethyl 4-(2-methoxyphenyl)-4-oxobutyrate

  • MDL number MFCD01311629
  • Cas Number : 103897-42-3
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7560B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7560B
Availability details for
Quantity
  • Availability
  • Add to Cart