RIE156330800 Rieke Metals, Inc.

6-oxoundecanoic acid

  • MDL number MFCD01320166
  • Cas Number : 91214-05-0
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7089D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7089D
Availability details for
Quantity
  • Availability
  • Add to Cart