RIE156330801 Rieke Metals, Inc.

7-Oxododecanoic acid

  • MDL number MFCD00786094
  • Cas Number : 54527-26-3
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7089E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7089E
Availability details for
Quantity
  • Availability
  • Add to Cart