RIE156330802 Rieke Metals, Inc.

ethyl 8-(2-methoxyphenyl)-8-oxooctanoate

  • MDL number MFCD01320261
  • Cas Number : 898752-76-6
  • Molecular Weight : 292.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7560F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7560F
Availability details for
Quantity
  • Availability
  • Add to Cart