RIE156330803 Rieke Metals, Inc.

Ethyl 4-(1-naphthyl)-4-oxobutyrate

  • MDL number MFCD01320262
  • Cas Number : 73931-66-5
  • Molecular Weight : 256.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7561B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7561B
Availability details for
Quantity
  • Availability
  • Add to Cart