RIE156330804 Rieke Metals, Inc.

4-Oxohexanoic acid

  • MDL number MFCD00068854
  • Cas Number : 1117-74-4
  • Molecular Weight : 130.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7087B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7087B
Availability details for
Quantity
  • Availability
  • Add to Cart