RIE156330805 Rieke Metals, Inc.

4-oxo-4-(4-n-propoxyphenyl)butyric acid

  • MDL number MFCD00100661
  • Cas Number : 39496-82-7
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7156B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7156B
Availability details for
Quantity
  • Availability
  • Add to Cart