RIE156330806 Rieke Metals, Inc.

4-(2,4,6-Trimethylphenyl)-4-oxobutyric acid

  • MDL number MFCD01096406
  • Cas Number : 15880-01-0
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7226B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7226B
Availability details for
Quantity
  • Availability
  • Add to Cart