RIE156330807 Rieke Metals, Inc.

5-(2,4,6-Trimethylphenyl)-5-oxovaleric acid

  • MDL number MFCD09801799
  • Cas Number : 500892-07-9
  • Molecular Weight : 234.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7226C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7226C
Availability details for
Quantity
  • Availability
  • Add to Cart