RIE156330808 Rieke Metals, Inc.

ethyl 4-(2-naphthyl)-4-oxobutyrate

  • MDL number MFCD01320266
  • Cas Number : 25370-42-7
  • Molecular Weight : 256.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7562B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7562B
Availability details for
Quantity
  • Availability
  • Add to Cart