RIE156330809 Rieke Metals, Inc.

4-(2,3,5,6-Tetramethylphenyl)-4-oxobutyric acid

  • MDL number MFCD09801800
  • Cas Number : 134948-69-9
  • Molecular Weight : 234.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7227B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7227B
Availability details for
Quantity
  • Availability
  • Add to Cart