RIE156330810 Rieke Metals, Inc.

5-(2,3,5,6-Tetramethylphenyl)-5-oxovaleric acid

  • MDL number MFCD02942028
  • Cas Number : 719295-43-9
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7227C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7227C
Availability details for
Quantity
  • Availability
  • Add to Cart