RIE156330811 Rieke Metals, Inc.

ethyl 7-(2-naphthyl)-7-oxoheptanoate

  • MDL number MFCD01311641
  • Cas Number : 898752-82-4
  • Molecular Weight : 298.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7562E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7562E
Availability details for
Quantity
  • Availability
  • Add to Cart