RIE156330812 Rieke Metals, Inc.

ethyl 8-(2-naphthyl)-8-oxooctanoate

  • MDL number MFCD01320269
  • Cas Number : 362669-46-3
  • Molecular Weight : 312.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7562F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7562F
Availability details for
Quantity
  • Availability
  • Add to Cart