RIE156330813 Rieke Metals, Inc.

4-(2,3,4,5,6-Pentamethylphenyl)-4-oxobutyric acid

  • MDL number MFCD00278208
  • Cas Number : 765269-95-2
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7228B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7228B
Availability details for
Quantity
  • Availability
  • Add to Cart