RIE156330814 Rieke Metals, Inc.

5-(2,3,4,5,6-Pentamethylphenyl)-5-oxovaleric acid

  • MDL number MFCD00118837
  • Cas Number : 655224-28-5
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7228C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7228C
Availability details for
Quantity
  • Availability
  • Add to Cart