RIE156330815 Rieke Metals, Inc.

4-(3,4-Dichlorophenyl)-4-oxobutyric acid

  • MDL number MFCD01464149
  • Cas Number : 50597-19-8
  • Molecular Weight : 247.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7120B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7120B
Availability details for
Quantity
  • Availability
  • Add to Cart