RIE156330816 Rieke Metals, Inc.

Ethyl 4-oxo-4-(4-trifluoromethylphenyl)butyrate

  • MDL number MFCD02261349
  • Cas Number : 155722-95-5
  • Molecular Weight : 274.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7641B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7641B
Availability details for
Quantity
  • Availability
  • Add to Cart