RIE156330818 Rieke Metals, Inc.

5-(4-Chloro-3-methylphenyl)-3-methyl-5-oxovaleric acid

  • MDL number MFCD06201678
  • Cas Number : 845781-51-3
  • Molecular Weight : 254.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7339

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7339
Availability details for
Quantity
  • Availability
  • Add to Cart