RIE156330820 Rieke Metals, Inc.

Ethyl 8-oxo-8-(4-trifluoromethylphenyl)octanoate

  • MDL number MFCD02261353
  • Cas Number : 362669-42-9
  • Molecular Weight : 330.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7641F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7641F
Availability details for
Quantity
  • Availability
  • Add to Cart