RIE156330822 Rieke Metals, Inc.

4-Oxo-4-(4-trifluoromethylphenyl)butyric acid

  • MDL number MFCD02260921
  • Cas Number : 58457-56-0
  • Molecular Weight : 246.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7141B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7141B
Availability details for
Quantity
  • Availability
  • Add to Cart