RIE156330823 Rieke Metals, Inc.

4-Oxo-4-(4-isopropoxyphenyl)butyric acid

  • MDL number MFCD02260991
  • Cas Number : 91121-67-4
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7157B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7157B
Availability details for
Quantity
  • Availability
  • Add to Cart