RIE156330824 Rieke Metals, Inc.

4-(5-Fluoro-2-methoxyphenyl)-4-oxobutyric acid

  • MDL number MFCD07790540
  • Cas Number : 49800-56-8
  • Molecular Weight : 226.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7232B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7232B
Availability details for
Quantity
  • Availability
  • Add to Cart