RIE156330828 Rieke Metals, Inc.

5-(2-Methoxy-5-methylphenyl)-5-oxovaleric acid

  • MDL number MFCD09801807
  • Cas Number : 71472-14-5
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7233C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7233C
Availability details for
Quantity
  • Availability
  • Add to Cart