RIE156330829 Rieke Metals, Inc.

ethyl 8-(4-bromophenyl)-8-oxooctanoate

  • MDL number MFCD01320180
  • Cas Number : 362669-43-0
  • Molecular Weight : 341.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7501F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7501F
Availability details for
Quantity
  • Availability
  • Add to Cart