RIE156330831 Rieke Metals, Inc.

5-(3-Chloro-5-fluorophenyl)-5-oxovaleric acid

  • MDL number MFCD06201696
  • Cas Number : 845790-48-9
  • Molecular Weight : 244.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7234C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7234C
Availability details for
Quantity
  • Availability
  • Add to Cart