RIE156330833 Rieke Metals, Inc.

5-(3-Fluoro-4-methoxyphenyl)-5-oxovaleric acid

  • MDL number MFCD06201658
  • Cas Number : 845781-33-1
  • Molecular Weight : 240.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7235C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7235C
Availability details for
Quantity
  • Availability
  • Add to Cart