RIE156330835 Rieke Metals, Inc.

Ethyl 4-(4-fluorophenyl)-4-oxobutyrate

  • MDL number MFCD01320182
  • Cas Number : 41310-80-9
  • Molecular Weight : 224.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7503B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7503B
Availability details for
Quantity
  • Availability
  • Add to Cart